missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: M-PA-C Agar

Catalog No. B98153
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B98153 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B98153 Supplier BD Supplier No. 298153

For the selective recovery of Pseudomonas auruginosa from water

Specifications

Certifications/Compliance SMWW
Form Dehydrated
Media Type M-PA-C Agar
Product Type Agar
Formulation Yeast extract 2g/L, L-Lysine HCl 5g/L, Sodium Chloride 5g/L, Xylose 1.25g/L, Sucrose 1.25g/L, Lactose 1.25g/L, Phenol red 0.08g/L, Ferric Ammonium Citrate 0.8g/L, Sodium Thiosulfate 5g/L, Magnesium Sulfate 1.5g/L, Kanamycin 8mg/L, Nalidixic Acid 37mg/L
For Use With (Application) Selective recovery of Pseudomonas auruginosa from Water
Packaging Type Bottle
Quantity 500 g