Learn More
BD BBL™ Dehydrated Culture Media: LBS (Lactobacillus Selection) Agar
For selective isolation and enumeration of lactobacilli
Supplier:  BD 211327
Catalog No. B11327
Only null left
Explore available promotions
Description
Description
Certifications
CE, COMPF, SMD
Specifications
Specifications
CE, COMPF, SMD
Dehydrated
Agar
For selective isolation and enumeration of lactobacilli
500 g
2°C to 8°C
LBS Agar
Pancreatic digest of Casein 10g/L, Yeast extract 5g/L, Monopotassium Phosphate 6g/L, Ammonium Citrate 2g/L, Dextrose 20g/L, Polysorbate 80 1g/L, Sodium Acetate Hydrate 25g/L, Magnesium Sulfate 575mg/L, Manganese Sulfate 0.12g/L, Ferrous Sulfate 34mg/L, Agar 15g/L
Bottle
Product Suggestions
Product Suggestions