missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: Eosin Methylene Blue Agar, Levine

Catalog No. B11221
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11221 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11221 Supplier BD Supplier No. 211221
Edge

Isolation of gram negative enteric bacteria

  • Formulation: Pancreatic digest of Gelatin 10g/L, Lactose 10g/L, Dipotassium Phosphate 2g/L, Eosin Y 0.4g/L, Methylene blue 65mg/L, Agar 15g/L

Certifications

AOAC, BAM, BS12, CCAM, CMPH2, COMPF, MCM9, SMD

Specifications

Certifications/Compliance AOAC, BAM, BS12, CCAM, CMPH2, COMPF, MCM9, SMD
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type EMB Agar, Levine
Product Type Agar
Formulation Pancreatic digest of Gelatin 10g/L, Lactose 10g/L, Dipotassium Phosphate 2g/L, Eosin Y 0.4g/L, Methylene blue 65mg/L, Agar 15g/L
For Use With (Application) Isolation of gram negative enteric bacteria
Packaging Type Bottle
Quantity 500 g