missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Columbia Agar Base
Specifications
Specifications
| Certifications/Compliance | AOAC, COMPF, EP, ISO, JP, USP |
| Content And Storage | 2 to 25°C |
| Form | Dehydrated |
| Media Type | Columbia Agar Base |
| Product Type | Agar |
| Formulation | Pancreatic digest of Casein 10g/L, meat Peptic digest 5g/L, Yeast extract 5g/L, heart Pancreatic digest 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 13.5g/L |
| For Use With (Application) | Growth of fastidious microbes |
| Packaging Type | Bottle |
| Quantity | 500 g |