missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: Brucella Broth

Catalog No. B11088
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11088 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11088 Supplier BD Supplier No. 211088
Edge

Dehydrated form for the convenience of the laboratorian and bioprocessing specialist. BD Difco™ BBL™ Dehydrated Culture Media: Brucella Broth is part of a large selection of dehydrated media.

  • Dehydrated
  • Culture Media
  • Brucella Broth

Specifications

Certifications/Compliance CCAM, ISO, USDA
Form Dehydrated
Media Type Brucella Broth
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Dextrose 1g/L, Yeast extract 2g/L, Sodium Chloride 5g/L, Sodium Bisulfite 0.1g/L
For Use With (Application) Cultivation of Brucella organisms
Packaging Type Bottle
Quantity 500 g