missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: Brain Heart Infusion Agar

Catalog No. B11065
Change view
Click to view available options
Brain Heart Infusion Agar; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
B11065 Brain Heart Infusion Agar; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11065 Supplier BD Supplier No. 211065
Edge

Use for isolation and cultivation of wide variety of fungal species, icluding bacteria, yeast and molds

 

  • Formulation: Brain heart infusion from (solids) 8g/L, Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 16g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L, Agar 13.5g/L

 

Certifications

AOAC, BAM, CCAM, COMPF, EPA, MCM9, SMD, SMWW, USDA

Specifications

Form Dehydrated
Media Type BHI Agar
Product Type Agar
Formulation Brain heart infusion from (solids) 8.0g/L, Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 16g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L, Agar 13.5g/L
For Use With (Application) For bacteria, Yeasts, molds and fungal species
Packaging Type Bottle
Quantity 500 g