missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: Blood Agar Base (Infusion Agar)

Catalog No. B11037
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11037 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11037 Supplier BD Supplier No. 211037
Only null left

For Streptococci and other fastidious

Specifications

Form Dehydrated
Media Type Blood Agar Base
Product Type Agar
Formulation Heart Muscle infusion from (solids) 2g/L, Pancreatic digest of Casein 13g/L, Yeast extract 5g/L, Sodium Chloride 5g/L, Agar 15g/L
For Use With (Application) For Streptococci and other fastidious
Packaging Type Bottle
Quantity 500 g