missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: BCYE Agar Base

Catalog No. B12327
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12327 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B12327 Supplier BD Supplier No. 212327

For isolation of Legionella species from clinical specimens and nonclinical (environmental) samples

  • Formulation: Yeast extract 10g/L, Ferric Pyrophosphate 0.25g/L, ACES Buffer 10g/L, Charcoal, Activated 2g/L, Alpha-Ketoglutarate 1g/L, Agar 15g/L

Certifications

SMWW

Specifications

Certifications/Compliance SMWW
Form Dehydrated
Media Type BCYE Agar base
Product Type Agar
Formulation Yeast extract 10g/L, Ferric Pyrophosphate 0.25g/L, ACES Buffer 10g/L, Charcoal, Activated 2g/L, Alpha-Ketoglutarate 1g/L, Agar 15g/L
For Use With (Application) For Legionella species
Packaging Type Bottle
Quantity 500 g