Learn More
BD BBL™ CultureSwab™ Plus Transport Systems: Amies Gel without Charcoal, Double Swab
For collection and transport of aerobes
Supplier:  BD 220117
Catalog No. B4320117
Only null left
Includes: Round-bottom tube; nontoxic rayon-tipped swab with 5.25 in. (13.3cm) shaft; security cap for a water-tight seal; extra-large, preattached label printed with lot number, expiration date and product description (also printed on peel pack); and tamper-evident seal
Description
Description
  • Provides enhanced performance for microorganism recovery
Specifications
Specifications
Collection and Transport System
Dual Applicator
Round bottom tube, nontoxic rayon tipped swab with 5.25 in. shaft, security cap for a water tight seal, extra large, preattached label printed with lot number, expiration date and product description and tamper evident seal
13.3 cm
Plastic
Amies Gel
Collection, transport and preservation of clinical specimens for Bacteriological examination
5.25 in.
50/Pk.
Rayon
Product Suggestions
Product Suggestions