missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ CHROMagar™ Orientation and TSA II I-Plate™

Catalog No. B222239
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
B222239 ISO/DIS, 11014
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B222239 Supplier BD Supplier No. 222239

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Two media in one plate to help streamline diagnosis

BBL CHROMagar Orientation and BBL Trypticase Soy Agar with 5% Sheep Blood (TSA II) I-Plate

BBL™ CHROMagar™ Orientation medium

  • Nonselective differentiated medium for isolation, differentiation and enumeration of urinary tract pathogens
  • Allows for differentiation and identification of Escherichia coli and Enterococcus without confirmatory testing
BBL™ Trypticase™ Soy Agar with 5% Sheep Blood
  • Used for cultivating fastidious microorganisms
  • Used for visualization of hemolytic reactions produced by many bacterial species

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Content And Storage 2°C to 8°C
Additive 5% Sheep Blood
Detects Urinary tract pathogens, Escherichia coli, Enterococcus, KES group, PMP group, Streptococcus pyogenes, Streptococcus pneumoniae, Staphylococcus aureus
Form Liquid
Manufacturing Origin United States
Media Type Chromagar™ orientation and Trypticase™ Soy Agar (TSA II) I-Plate
Formulation Chromopeptone 16.1g/L, Chromogen Mix 1.3g/L, Agar 15g/L, Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, Growth factors 1.5g/L, Defibrinated Sheep Blood 5%
For Use With (Application) Urinary tract pathogens, Escherichia coli and Enterococcus
Includes Chromagar™ orientation medium on one side and Soy Agar with 5% Sheep Blood (TSA II)-I medium on the other side
Incubation Conditions 35 ±2 °C, Aerobic
Incubation Time 18 to 24 hr.
Packaging Type Split Plate
Quantity 100/Pk.
Show More Show Less