missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ CDC Anaerobic Blood Agar Sterile Pack Media - Rodac™ Plates
Description
Rodac (Replicate Organism Detection and Counting) plates for surface sampling.
To use:
- Place wrapped package of plates in the isolator before running the vaporized hydrogen peroxide decontamination cycle
- When the isolator aeration cycle has finished, remove the two outermost wrappings, then the sterile inner wrapping
- Use the included inner bag to transport plates from your critical environment
Compliance
Tested for performance and sterility according to Association for the Advancement of Medical Instrumentation (AAMI) guidelines.
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Yellow |
| Content And Storage | 2°C to 8°C |
| Format | Rodac |
| Manufacturing Origin | United States |
| Media Type | CDC anaerobic Blood Agar |
| Sterility | Sterile |
| For Use With (Application) | Surface and active air sampling |
| Includes | Inner bag for transportation |
| Packaging Type | Plate |
| Show More |