Learn More
BD BBL™ AFB Slides
Catalog No. B231391
Change view
Click to view available options
Quantity:
50 Pk.
Catalog No. Quantity
B231391 50 Pk.
1 options
Catalog No. B231391 Supplier BD Supplier No. 231391
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For evaluating and controlling the quality of acid-fast and fluorescent stain reagents and staining techniques for the detection of acid-fast bacilli (AFB)
Specifications
Product Type Control Slide
Quantity 50 Pk.
For in vitro diagnostic use.