missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Bacto™ Dehydrated Culture Media: Tryptic Soy Broth without Dextrose
Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms
Supplier: BD 286220
Description
Cultivation of fastidious and non-fastidious microorganisms from variety of clinical and nonclinical specimens
Specifications
| BAM | |
| Dehydrated | |
| Microbiology Media | |
| Cultivation of fastidious and non-fastidious microorganisms from a variety of clinical and nonclinical specimens | |
| 500 g |
| 2°C to 8°C | |
| Tryptic Soy Broth | |
| Pancreatic digest of Casein 17g/L, Enzymatic digest of Soybean Meal 3g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L | |
| Bottle |