missing translation for 'onlineSavingsMsg'
Learn More

BD Bacto™ Dehydrated Culture Media: Todd Hewitt Broth

Catalog No. DF0492176
Change view
Click to view available options
Quantity:
500 g
Packaging Type:
Bottle
For Use With (Application):
Hemolytic Streptococci growth
Form:
Dehydrated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type For Use With (Application) Form
DF0492176 500 g Bottle Hemolytic Streptococci growth Dehydrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0492176 Supplier BD Supplier No. 249240
Edge

For cultivation of group A streptococci, pathogenic microorganisms or as a blood culture medium

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage <30°C
Form Dehydrated
Media Type Todd Hewitt Broth
Product Type Microbiology Media
Formulation Heart infusion from 500g 3.1g/L, Neopeptone 20g/L, Dextrose 2g/L, Sodium Chloride 2g/L, Disodium Phosphate 0.4g/L, Sodium Carbonate 2.5g/L
For Use With (Application) Hemolytic Streptococci growth
Packaging Type Bottle
Quantity 500 g