missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Brain Heart Infusion
GREENER_CHOICE

Catalog No. df0037089
Change view
Click to view available options
Quantity:
10 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0037089 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0037089 Supplier Gibco™ Supplier No. 237300

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used in the cultivation of fastidious and nonfastidious microorganisms. Gibco™ Bacto™ Brain Heart Infusion is a general-purpose medium used to cultivate aerobic and anaerobic bacteria from a variety of clinical and nonclinical materials.

BHI broth is used for the cultivation of a wide variety of microorganisms, including bacteria (streptococci, pneumococci and meningococci), yeasts and molds. It serves as a base for supplemented media containing 0.1% agar, Fildes enrichment or 6.5% sodium chloride.

  • To use dehydrated product: suspend 37g in 1L of purified water
  • Displays a good cultural response to Neisseria meningitides 13090, Streptococcus pneumonia 6305 and Streptococcus pyogenes 19615
  • pH (3.7% solution, 25°C): 7.4 ± 0.2

Certifications

AOAC, BAM, CCAM, CLSI, CMPH2, COMPF, EPA, ISO, MCM9, SMD, SMWW, USDA

Specifications

Certifications/Compliance AOAC, BAM, CCAM, CLSI, CMPH2, COMPF, EPA, ISO, MCM9, SMD, SMWW, USDA
Color Light tan
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Form Dehydrated (free-flowing, homogenous)
Media Type Brain Heart Infusion
Product Type Brain Heart Infusion
Formulation Calf brain (infusion from 200g) 7.7g/L, Beef heart (infusion from 250g) 9.8g/L, Proteose Peptone 10g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L
For Use With (Application) Cultivation of fastidious and non-fastidious microorganisms, including aerobic and anaerobic bateria, yeasts and molds
Packaging Type Pail
Product Line Bacto
Quantity 10 kg
Show More Show Less