Learn More
BD-10A High-Frequency Generator
Catalog No. S43504
Encompass
Change view
Click to view available options
Includes:
Standard electrode tip
Catalog No. Includes
S43504 Standard electrode tip
1 options
Catalog No. S43504 Supplier Electro Technic Products Supplier No. 11011
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Standard tester model.
  • 115V 50/60Hz with standard electrode tip
  • 10,000 to 50,000V 500KHz
  • No switch
Specifications
Voltage 115 V
Includes Standard electrode tip
Product Type High Frequency Generator