MW 372.9 Da, Purity >98%. Potent inhibitor of TRPV1 (IC values are 35 and 6 nM for capsaicin and acid-induced activation respectively). Inhibits TRPM8 (IC = 0.8 M). Reduces hyperalgesia in vivo..
The product is subject to the following: Abcam Restricted Use Statement