missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ BCR™ Single-Knit Nylon Full-Finger Cleanroom Glove Liners

Catalog No. p-7230585
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19037612A Small
19037612C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19037612A Supplier Berkshire™ Supplier No. BGL7.200SB
Only null left

Single-knit nylon construction is thin and comfortable, while the full-finger design provides total protection for those with severe glove sensitivities.

  • Reduces perspiration and contact dermatitis commonly associated with wearing latex, urethane, PVC, or nitrile gloves
  • Full-finger design provides total protection for those with severe glove sensitivities
  • Can be worn alone as inspection gloves to prevent fingerprints on critical surfaces
  • Single-knit nylon construction is thin and comfortable
  • Ambidextrous design can be used on either hand
  • Gloves can be repeatedly laundered for reuse
  • Ideal for wet or temperature related processing
  • Suitable for use in ISO Class Cleanrooms

Specifications

Construction Single Knit
Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Material Nylon
Size Small
Finger Style Full
For Use With (Application) Wet/Temperature-Related Processing, Preventing Contact Dermetitis and Perspiration
Glove Cuff Style Knit
Hem Color Green
Series BGL7
Show More Show Less