missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ BCR™ Polyester Half-Finger Cleanroom Glove Liners

Catalog No. p-7230588
Change view
Click to view available options
Size:
Regular
Large
Packaging Type:
Case
Pack
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Packaging Type
19037187 Regular Pack
19037599 Regular Case
19037188 Large Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19037187 Supplier Berkshire™ Supplier No. BGL2.20R
Only null left

Continuous filament knitted polyester construction is thin yet durable, while the half-finger design permits tactility in addition to wicking moisture from the palms.

  • Reduces perspiration and contact dermatitis commonly associated with wearing latex, urethane, PVC, or nitrile gloves
  • Half-finger design permits tactility while also wicking moisture from the palms
  • 100% continuous filament knitted polyester is thin, yet durable
  • Ambidextrous design can be used on either hand
  • Gloves can be repeatedly laundered for reuse
  • Suitable for use in ISO Class Cleanrooms

Specifications

Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Material Polyester
Size Regular
Finger Style Half
For Use With (Application) Laboratories and Manufacturing Environments
Glove Cuff Style Knit
Hem Color White
Packaging Type Pack
Series BGL2
Show More Show Less