Learn More
Bioassay Systems QuantiChrom™ BCP Albumin Assay Kit. For quantitative determination of albumin.
Catalog No. 50489209
Encompass
For quantitative determination of albumin and evaluation of drug effects on albumin metabolism. Key Features: Sensitive and accurate. Use as little as 20 µL samples. Detection range 0.3 - 5 g/dL (45 - 750 µM) albumin in 96-well plate assay. Simple and high-throughput. The procedure involves addition of a single working reagent and incubation for 5 min. Can be readily automated as a high-throughput assay for thousands of samples per day. Improved reagent stability and versatility. The optimized formulation has greatly enhanced reagent and signal stability. Cuvet or 96-well plate assay. Low interference in biological samples. No pretreatments are needed. Assays can be directly performed on raw biological samples i.e., in the presence of lipid and protein. Method: OD610nm (BCP). Samples: Serum, plasma, urine, biological preparations. Species: All. Procedure: Assay takes 5 min. Kit size: 250 tests. Detection limit: 0.3 g/dL.
Catalog No. 50489209 Supplier Bioassay Systems Supplier No. DIAP250
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For quantitative determination of albumin and evaluation of drug effects on albumin metabolism. Key Features: Sensitive and accurate. Use as little as 20 µL samples. Detection range 0.3 - 5 g/dL (45 - 750 µM) albumin in 96-well plate assay. Simple and high-throughput. The procedure involves addition of a single working reagent and incubation for 5 min. Can be readily automated as a high-throughput assay for thousands of samples per day. Improved reagent stability and versatility. The optimized formulation has greatly enhanced reagent and signal stability. Cuvet or 96-well plate assay. Low interference in biological samples. No pretreatments are needed. Assays can be directly performed on raw biological samples i.e., in the presence of lipid and protein. Method: OD610nm (BCP). Samples: Serum, plasma, urine, biological preparations. Species: All. Procedure: Assay takes 5 min. Kit size: 250 tests. Detection limit: 0.3 g/dL.