Learn More
ACROBiosystems Inc ACROBiosystems Mouse BCMA / TNFRSF17 Protein, His Tag
Catalog No. 501957225
Encompass
Recombinant Protein;1MG;Mouse BCMA, His Tag (BCA-M52H3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Thr 49 (Accession # O88472-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.3 kDa. The protein migrates as 10 kDa, 13 kDa and 15-16 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse BCMA, His Tag (Cat. No. BCA-M52H3) at 0.5 g/mL (100 L/well) can bind Mouse BAFF, Mouse IgG2a Fc Tag (Cat. No. BAF-M5257) with a linear range of 0.1-6 ng/mL (QC tested).;Immobilized Mouse BCMA, His Tag (Cat. No. BCA-M52H3) at 2 g/mL (100 L/well) can bind BCMA Antibody (Mouse IgG1, the antibody was co developed by SCT and ACRObiosystems) with a linear range of 0.256-6.4 ng/mL (Routinely tested).
Catalog No. 501957225 Supplier ACROBiosystems Inc Supplier No. BCAM52H31MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;1MG;Mouse BCMA, His Tag (BCA-M52H3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Thr 49 (Accession # O88472-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.3 kDa. The protein migrates as 10 kDa, 13 kDa and 15-16 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse BCMA, His Tag (Cat. No. BCA-M52H3) at 0.5 g/mL (100 L/well) can bind Mouse BAFF, Mouse IgG2a Fc Tag (Cat. No. BAF-M5257) with a linear range of 0.1-6 ng/mL (QC tested).;Immobilized Mouse BCMA, His Tag (Cat. No. BCA-M52H3) at 2 g/mL (100 L/well) can bind BCMA Antibody (Mouse IgG1, the antibody was co developed by SCT and ACRObiosystems) with a linear range of 0.256-6.4 ng/mL (Routinely tested).