Learn More
Abcam Inc BCIP disodium salt, histochemical alkaline phosphatase substrate, 100MG
Catalog No. 502704823
Encompass
MW 370.43 g/mol. Chromogenic substrate for alkaline phosphatase. Often used in combination with nitro blue tetrazolium chloride (NBT) to amplify the sensitivity of staining.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502704823 Supplier Abcam Inc Supplier No. AB146226100MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 370.43 g/mol. Chromogenic substrate for alkaline phosphatase. Often used in combination with nitro blue tetrazolium chloride (NBT) to amplify the sensitivity of staining.
The product is subject to the following: Abcam Restricted Use Statement