missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ BCECF/AM Calbiochem™,

Catalog No. 2162541MG
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2162541MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2162541MG Supplier MilliporeSigma™ Supplier No. 2162541MG
Only null left

A variable mixture of cell-permeable ester derivatives of BCECF that are hydrolyzed by cytosolic esterases to yield intracellularly trapped indicator BCECF

Chemical Identifiers

CAS 117464-70-7
Molecular Formula C42H40O21
Molecular Weight (g/mol) 880.76
MDL Number MFCD00036969
InChI Key NTECHUXHORNEGZ-UHFFFAOYSA-N
Synonym bcecf-acetoxymethyl, bcecf-am solution, 2',7'-bis 2-carboxyethyl-5 6-carboxyfluorescein acetoxymethyl ester, acetyloxy methyl 3-3',6'-bis acetyloxy methoxy-2'-3-acetyloxy methoxy-3-oxopropyl-5-acetyloxy methoxy carbonyl-3-oxospiro 2-benzofuran-1,9'-xanthen-7'-yl propanoate, bcecf-am solution 5 mm in dmso , 1 mg in 0.25 ml dmso, spiro isobenzofuran-1 3h ,9'-9h xanthene-2',7'-dipropanoic acid, 2',7'-bis 2-carboxyethyl-5 6-carboxyfluorescein acetoxymethyl ester mixed isomers, 2',7'-bis 2-carboxyethyl-5 6-carboxyfluorescein tetrakis-acetoxymethyl ester, 2 inverted exclamation marka,7 inverted exclamation marka-bis 2-carboxyethyl-5 6-carboxyfluorescein acetoxymethyl ester, 2 inverted exclamation marka,7 inverted exclamation marka-bis 2-carboxyethyl-5 6-carboxyfluorescein acetoxymethyl ester mixed isomers
PubChem CID 53229972
IUPAC Name (acetyloxy)methyl 3-{3',6'-bis[(acetyloxy)methoxy]-7'-{3-[(acetyloxy)methoxy]-3-oxopropyl}-5-{[(acetyloxy)methoxy]carbonyl}-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-2'-yl}propanoate
SMILES CC(=O)OCOC(=O)CCC1=C(OCOC(C)=O)C=C2OC3=CC(OCOC(C)=O)=C(CCC(=O)OCOC(C)=O)C=C3C3(OC(=O)C4=C3C=CC(=C4)C(=O)OCOC(C)=O)C2=C1

Specifications

Color White
Quantity 1 mg
Percent Purity ≥93%
Grade HPLC
Physical Form Solid
Chemical Name or Material BCECF/AM
Recommended Storage -20°C (-4°F); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above -20°C (-4°F).