Learn More
Thermo Scientific Chemicals BCA Protein Assay Kit
Catalog No. aaj63283qa
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
AAJ63283QA 1 Kit
1 options
Catalog No. AAJ63283QA Supplier Thermo Scientific Chemicals Supplier No. J63283QA
Only null left
BCA Protein Assay Kit can be used to measure very dilute protein concentrations in very small sample volumes. Accurately measures protein concentrations from 0.5 to 30 ?g/mL in tube assays and 1 to 20 ?g/mL in 96- or 384-well plate assays. It is used for studying protein: protein interactions, measuring column fractions after affinity chromatography, estimating percent recovery of membrane proteins from cell extracts and high-throughput screening of fusion proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Description Clear
Product Type Protein Assay Kit
For Use With (Application) Contains two separate solutions for estimation of protein concentration.
Quantity 1 Kit
RUO – Research Use Only