Learn More
Brady™ BBP31 Hi-Performance Polyester Tapes - Brown
For labelling stainless steel and smooth surfaces. Brady™ BBP31 Hi-Performance Polyester Tapes are made of B-569 polyester and are ideal for application to very smooth surfaces or rigid panels.
Supplier:  Brady™ B30C2250569BR
 View more versions of this product
Catalog No. 19108053
Description
Description
  • Applied to very smooth surfaces
  • Ideal for mounting to rigid panels
Specifications
Chemical Identifiers
Compatible surface: Smooth
Brown
Cartridge
Marking stainless steel pipes
1
Brady Workstation, LabelMark, MarkWare
0.003 in.
100 ft.
Acrylic, Permanent
Gloss
Polyester (B-569)
BBP30, BBP31, BBP33, BBP35, BBP37 printers
1 Ea.
-40°C to +100°C
0.076 mm
30.40 m
Specifications
Compatible surface: Smooth
Brown
Cartridge
BBP31 Hi-Performance Polyester Tape
BBP30, BBP31, BBP33, BBP35, BBP37 printers
1 Ea.
-40°C to +100°C
0.076 mm
30.40 m
2.25 in.
Acrylic, Permanent
Gloss
Polyester (B-569)
Marking stainless steel pipes
1
Brady Workstation, LabelMark, MarkWare
0.003 in.
100 ft.
57 mm
Product Suggestions
Product Suggestions