missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Streptocard™ Acid Latex Test Kit

Catalog No. B240917
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B240917 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B240917 Supplier BD Supplier No. 240917

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Streptocard Acid Test Latex A, B, C, D, F, and G Reagents, 2.5mL each; Streptocard Acid Positive Control, 2.5mL; Streptocard Acid Extraction Reagents 1 and 2, 2.7mL each; Streptocard Acid Extraction Reagent 3, 2 x 6.8mL; Test cards; Mixing sticks

Provides a rapid platform for the serological identification of beta-haemolytic streptococci belonging to Lancefield groups A, B, C, D, F and G

Beta- haemolytic streptococci are the most frequently isolated human pathogens among the representatives of the genus Streptococcus. Nearly all the beta-haemolytic streptococci possess specific carbohydrate antigens (streptococcal group antigens). Lancefield showed that these antigens can be extracted in soluble form and identified by precipitation reactions with homologous antisera. The BD BBL Streptocard Acid Latex Test Kit is based on liberation of specific antigen from bacteria cell walls by modified nitrous acid extraction. The extracted antigen in conjunction with latex agglutination offers a rapid, sensitive and specific method for identification of streptococcal groups A, B, C, D, F and G from primary culture plates.

Specifications

Content And Storage 2°C to 8°C Do not freeze
Description Streptocard Acid Latex Test Kit
Detectable Analytes beta-haemolytic Streptococci (Groups A, B, C, F, G)
For Use With (Application) Streptococcus Testing
Includes Streptocard Acid Test Latex A, B, C, D, F, and G Reagents, 2.5mL each; Streptocard Acid Positive Control, 2.5mL; Streptocard Acid Extraction Reagents 1 and 2, 2.7mL each; Streptocard Acid Extraction Reagent 3, 2 x 6.8mL; Test cards; Mixing sticks
Quantity 50 Tests

For in vitro diagnostic use.