Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs for Veterinary Laboratory Use
Same as standard Sensi-Disc Susceptibility Test Discs, but for veterinary laboratory use.
Supplier:  BD 231717
 View more versions of this product
Catalog No. B4331717
Only null left
Explore available promotions
Description
Description
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents.
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution
  • Clearly marked on both sides with letters and numbers designating agent and concentration
Specifications
Specifications
Enrofloxacin
5 μg
Disc
50 Discs
Product Suggestions
Product Suggestions