Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs for Veterinary Laboratory Use
Catalog No. B4391255
Change view
Click to view available options
Concentration:
30 μg
5 μg
Antimicrobial Agents:
Ceftiofur
Enrofloxacin
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration Antimicrobial Agents
B4391255 30 μg Ceftiofur
B4331717 5 μg Enrofloxacin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4391255 Supplier BD Supplier No. 291255
Same as standard Sensi-Disc Susceptibility Test Discs, but for veterinary laboratory use.
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents.
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution
  • Clearly marked on both sides with letters and numbers designating agent and concentration
Specifications
Antimicrobial Agents Ceftiofur
Concentration 30 μg
Packaging Type Disc
Quantity 50 Discs