missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Ceftazidime/Avibactam (CZA50)

Catalog No. B232218
Change view
Click to view available options
Concentration:
30/20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
B232218 30/20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B232218 Supplier BD Supplier No. 232218
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Employs the Bauer-Kirby disc diffusion testing method.

  • Flexibility in testing
  • Continuous development of discs containing the latest antimicrobics
  • Large VFD is clearly visible even in poorly-lit workplace
  • Antimicrobial agents impregnated on high-quality paper
  • 50 discs in each cartridge
  • Self-tamping dispensers: single, 6, 8 or 12 place

Specifications

Antimicrobial Agents Ceftazidime/Avibactam
Concentration 30/20 μg
Form Solid
Product Type Antimicrobial Susceptibility Discs
Packaging Type Disc