missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Media: Trypticase™ Soy Agar (TSA II™) with Sheep Blood
Used for cultivating fastidious microorganisms and for visualization of hemolytic reactions produced by many bacterial species
$208.07 - $208.07
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
|---|---|
| Content And Storage | 2°C to 8°C |
| Manufacturing Origin | United States |
| Formulation | Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, Growth Factors 1.5g/L, Defibrinated Sheep Blood 5% or 10% |
| For Use With (Application) | Cultivation of fastidious microorganisms and visualization of hemolytic reactions |
Description
- Ready for immediate use
- Available with 5% and 10% sheep blood
- “Deep-fill labelled plates contain additional volume of medium to reduce effects of drying during prolonged incubation”
Specifications
| ISO/DIS, 11014 | |
| United States | |
| Cultivation of fastidious microorganisms and visualization of hemolytic reactions |
| 2°C to 8°C | |
| Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, Growth Factors 1.5g/L, Defibrinated Sheep Blood 5% or 10% | |
| Plate |