missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Prepared Plated Media: Trypticase™ Soy Agar (TSA II™) with Sheep Blood

Catalog No. B21261
Change view
Click to view available options
Color:
Dark Red
Media Type:
TSA II with 5% Sheep Blood
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Media Type
B21261 Dark Red TSA II with 5% Sheep Blood
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B21261 Supplier BD Supplier No. 221261

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for cultivating fastidious microorganisms and for visualization of hemolytic reactions produced by many bacterial species

  • Ready for immediate use
  • Available with 5% and 10% sheep blood
  • “Deep-fill labelled plates contain additional volume of medium to reduce effects of drying during prolonged incubation”

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Dark Red
Content And Storage 2°C to 8°C
Format Plate
Manufacturing Origin United States
Media Type TSA II with 5% Sheep Blood
Product Type Prepared Microbiology Plate
Target Organism Class Bacteria
Test Type Microbial Cultivation
Formulation Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, Growth Factors 1.5g/L, Defibrinated Sheep Blood 5% or 10%
For Use With (Application) Cultivation of fastidious microorganisms and visualization of hemolytic reactions
Incubation Conditions 35 ±2°C
Incubation Time 18 to 24 hr.
Packaging Type Plate
Quantity 100/Pk.
Height (Metric) 15 mm
Protein Source Casein, Soy
Show More Show Less