missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Prepared Plated Media: Columbia Agar with 5% Sheep Blood

Catalog No. p-4766065
Change view
Click to view available options
Quantity:
100/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21263 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B21263 Supplier BD Supplier No. 221263
Only null left

For the isolation of microorganisms from samples or specimens

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Red
Form Solid
Manufacturing Origin United States
Media Type Columbia Agar with 5% Sheep Blood
Formulation Pancreatic digest of Casein 12g/L, Peptic digest of Animal Tissue 5g/L, Yeast extract 3.5g/L, Beef extract 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Sheep Blood, Defibrinated 5%
For Use With (Application) Non-fastidious and fastidious microorganisms
Packaging Type Plate
Quantity 100/Pk.