missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Media: Columbia Agar with 5% Sheep Blood
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Red |
| Form | Solid |
| Manufacturing Origin | United States |
| Media Type | Columbia Agar with 5% Sheep Blood |
| Formulation | Pancreatic digest of Casein 12g/L, Peptic digest of Animal Tissue 5g/L, Yeast extract 3.5g/L, Beef extract 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Sheep Blood, Defibrinated 5% |
| For Use With (Application) | Non-fastidious and fastidious microorganisms |
| Packaging Type | Plate |
| Quantity | 100/Pk. |