missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Media: CHROMagar™ MRSA II
Description
- BD BBL™ CHROMagar™ MRSA II is a selective, differential medium for qualitative, direct identification of methicillin-resistant Staphylococcus aureus (MRSA).
- It is suitable for direct detection of nasal colonization by MRSA from anterior nares specimens.
- Validated for use with the BD Kiestra™ MRSA imaging application for automated growth detection and color recognition to enable mauve color growth detection without confirmatory testing for nares specimens only.
- Validated for use with the BD Kiestra™ MRSA imaging application for automated growth detection and color recognition to enable mauve color growth detection without confirmatory testing for nares specimens only.
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Amber |
| Content And Storage | 2°C to 8°C |
| Detects | Methicillin-resistant Staphylococcus aureus |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Chromagar™ MRSA II |
| Test Type | Microbial Detection |
| Formulation | Chromopeptone 35g/L, Chromogen Mix 0.5g/L, Sodium Chloride 17.5g/L, Inhibitory Agents 7.5g/L, Cefoxitin 5.2g/L, Agar 14g/L |
| For Use With (Application) | Selective and differential chromogenic medium for the qualitative direct detection of methicillin resistant Staphylococcus aureus from anterior nares specimens |
| Show More |