missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Prepared Plated Media: CDC Anaerobe 5% Sheep Blood Agar

For the isolation and cultivation of fastidious and slow growing, obligately anaerobic bacteria from a variety of clinical and nonclinical materials

$49.34 - $49.34

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Form Liquid
Manufacturing Origin United States
Media Type CDC anaerobe 5% Sheep Blood Agar
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B21734X
View Documents
BD
221734
100/Pk.
Pack of 100 Each for $49.34
Only null left
 
Description

Description

It also supports the growth of most aerobic, facultatively anaerobic and microaerophilic bacteria if incubated appropriately.

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

Specifications

Specifications

ISO/DIS, 11014
Liquid
CDC anaerobe 5% Sheep Blood Agar
Plate
Light Beige
United States
Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 20g/L, Yeast extract 5g/L, Hemin 0.005g/L, Vitamin K1 0.01g/L, L-Cystine 0.4g/L, Sheep Blood, defibrinated 5%
950 g
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'