missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Prepared Plated Chromogenic Media: MI Agar

Catalog No. B14985
Change view
Click to view available options
Quantity:
100/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14985 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B14985 Supplier BD Supplier No. 214985
Only null left

Culture media tests drinking water for the presence of E.coli and total coliforms with membrane filtration method

Detects degree of water quality deterioration in drinking distribution system enumerates, and monitors the progress in remediation. Provides precise and quantifiable results; also less time consuming and costly than other techniques.

  • Chromogenic-fluorogenic medium for detecting total coliforms and Escherichia coli in 24 hours or less
  • Under ambient light, E. coli exhibits bluish-gray appearance; under long-wave UV light, it displays blue-green fluorescence
  • Coliform are cream under ambient light, and display blue-white fluorescence under long-wave UV light
  • Conforms to EPA Method 1604 for monitoring drinking water under the Total Coliform Rule, and source water under the Surface Water Treatment

Applications:

For testing ambient and potable water, treatment plant effluents, and beverage and pharmaceutical manufacturing

Order Info

Available dehydrated or in prepared 15 x 60mm plates.

Specifications

Certifications/Compliance US, EPA
Color Beige
Content And Storage 25°C
Format Prepared Plate
Manufacturing Origin United States
Media Type MI Agar
Formulation No. 3 Proteose Peptone 5g/L, Yeast extract 3g/L, Lactose 1g/L, 4-Methylumbelliferyl-beta-D-Galactopyranoside 0.1g/L, Indoxyl-beta-D-Glucuronide 0.32g/L, Sodium Chloride 7.5g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 1g/L, Sodium Lauryl Sulfate 0.2g/L, Sodium Desoxycholate 0.1g/L, Agar 15g/L, Cefsulodin 5mg/L
For Use With (Application) To detect and enumerate Escherichia coli and total Coliforms in drinking Water by the membrane filtration technique
Packaging Type Plate
Quantity 100/Pk.