missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Mycoflask™ Trypticase™ Soy Broth Prepared Media
Used for the cultivation of fastidious and nonfastidious microorganisms from a variety of clinical and nonclinical specimens
Supplier: BD 221715
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
| CE, AOAC, BAM, BS12, CCAM, CLSI, CMPH2, COMPF, EP, EPA, ISO, JP, MCM9, SMD, USDA, USP | |
| 15°C to 25°C | |
| United States | |
| Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Dextrose 2.5g/L | |
| Size K tubes with cap |
| Beige | |
| Liquid | |
| Trypticase Soy Broth | |
| For qualitative procedures for the cultivation of fastidious and non-fastidious microorganisms | |
| 10 x 5 mL |
For in vitro diagnostic use.