Learn More
BD BBL™ Gram Stain Kits
Catalog No. B4312539
Change view
Click to view available options
Type:
Gram Stain Kit (Stabilized)
Catalog No. Type
B4312539 Gram Stain Kit (Stabilized)
1 options
Catalog No. B4312539 Supplier BD Supplier No. 212539
Only null left
Used to stain microorganisms from cultures or specimens by the differential Gram method
  • Complete stain kits reduce the mess associated with Gram staining procedures
  • Each break-resistant solution bottle features a hinged spot cap that makes regulating the flow easy and helps prevent spots and spills Color-coded labels allow quick identification of reagents
  • Stabilized Iodine Solution requires no mixing and has a long shelf life
Specifications
Type Gram Stain Kit (Stabilized)
Quantity 1 Kit
Certifications/Compliance CE
Includes 250mL gram crystal violet, 250mL iodine, 250mL gram decolorizer and 250mL gram safranin
For Use With (Application) Used to stain microorganisms from cultures or specimens by the differential gram method
Stain Type Gram Stain