missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Transport Medium (Stuart, Toshach and Patsula)
Used for collecting, transporting and preserving microbiological specimens
Supplier: BD 211743
Specifications
| CE | |
| Dehydrated | |
| Microbiology Media | |
| For collecting, transporting and preserving microbiological specimen | |
| 500 g |
| 2°C to 8°C | |
| Transport Medium (Stuart, Toshach and Patsula) | |
| Sodium Thioglycollate 1g/L, Sodium Glycerophosphate 10g/L, Calcium Chloride 0.1g/L, Methylene blue 2mg/L, Agar 3g/L | |
| Bottle |