Learn More
BD BBL™ Crystal™ ID Systems Accessories
Catalog No. p-44609
Change view
Click to view available options
Product Type:
Calibration Standard
Panel Viewer
Quantity:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
B4345015 Calibration Standard Each
1491069 Panel Viewer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4345015 Supplier BD Supplier No. 245015
Only null left
BBL Crystal MIND Software
  • Interprets the test results and provides specimen identification
  • Minimum software requirements are an MS-DOS™ 5.0 or higher compatible computer; Intel™ 486 microprocessor with at least 16MB RAM Software is compatible with Microsoft™ Windows™ (3.11, 95, 98, 2000, XP) Computer must have at least Microsoft ODBC Version 2.00 and a 3.5 in. floppy drive
Codebooks Include Enteric/Nonfermenter, Anaerobe, Neisseria/Haemophilus, Gram-Positive, Rapid Gram-Positive) Supplied in individual hard-copy binder BBL Crystal Panel Viewer
  • For facilitated viewing and recording of chromogenic and fluorogenic reactions
BBL CrystalSpec™ Standards†
  • Set of two calibration standards for the CrystalSpec Nephelometer, one Calibration Blank and one Calibration Standard
†Drop shipment is required. All shipments FOB from Cockeysville, MD. Freight charges will be prepaid and billed as a separate item on the invoice.
Specifications
Product Type Calibration Standard
For Use With (Equipment) CrystalSpec™ nephelometer
Includes Set of two calibration standard, one calibration blank and one calibration standard
Quantity Each