Learn More
Eisco™ Battery Snap Connector, 9 Volt
Battery Snap Connector for use with 9 Volt - I Type Batteries
Supplier:  Eisco™ PH0953E
Catalog No. S27754
Only null left
Explore available promotions
Description
Description
  • Snap-on connector with insulated leads
  • Fits 9-Volt, I Type Batteries
  • Red and black leads
  • Each lead measures 6 in. long
Specifications
Specifications
Battery Snap Connector
Insulated Leads
1 Ea.
9 Volt
Product Suggestions
Product Suggestions