missing translation for 'onlineSavingsMsg'
Learn More

Bathophenanthrolinedisulfonic acid disodium salt trihydrate, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. p-7187994
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014465 1 g
111014464 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014465 Supplier Merck Emd Millipore Supplier No. 118901G

Chemical Identifiers

Synonym 4,7-Diphenyl-1,10-phenanthroline-disulfonic acid disodium salt trihydrate; Disodium bathophenanthrolinedisulfonate trihydrate

Specifications

CAS 52746-49-3
Linear Formula C24H14N2 Na2O6S2 · 3H2 O
Quantity 1 g
Formula Weight 590.53
Percent Purity ≥98.0% (HPLC)