missing translation for 'onlineSavingsMsg'
Learn More

Bathophenanthrolinedisulfonic Acid Disodium Salt Hydrate [for Determination of Ferrous Ion], TCI America™

Catalog No. B09891G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B09891G Supplier TCI America Supplier No. B09891G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 53744-42-6
Molecular Formula C24H14N2Na2O6S2
Molecular Weight (g/mol) 536.484
MDL Number MFCD00038854
InChI Key PCNDSIWXTYFWIA-UHFFFAOYSA-L
PubChem CID 104311
ChEBI CHEBI:78157
IUPAC Name disodium;4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzenesulfonate
SMILES C1=CC(=CC=C1C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Red-White
Quantity 1 g
Formula Weight 536.48
Physical Form Crystalline Powder
Chemical Name or Material Bathophenanthrolinedisulfonic Acid Disodium Salt Hydrate [for Determination of Ferrous Ion]