missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bathophenanthroline 99.0+%, TCI America™
Supplier: TCI America D09055G
Specifications
| Bathophenanthroline | |
| 221°C | |
| C24H16N2 | |
| 5 g | |
| DHDHJYNTEFLIHY-UHFFFAOYSA-N | |
| 4,7-diphenyl-1,10-phenanthroline | |
| 72812 | |
| 332.41 | |
| Crystalline Powder |
| 1662-01-7 | |
| White-Yellow | |
| MFCD00004976 | |
| bathophenanthroline, 1,10-phenanthroline, 4,7-diphenyl, bphen, 1,10-bathophenanthroline, bathophenanthrolin, bathophenanthrolin german, unii-4a2b091f0g, 4,7-diphenyl-1,10-diazaphenanthrene, gnf-pf-4554, 4,7-diphenylpyridino 3,2-h quinoline | |
| C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5 | |
| 332.406 | |
| CHEBI:77995 | |
| ≥99.0% (T) |
Chemical Identifiers
| 1662-01-7 | |
| 332.406 | |
| DHDHJYNTEFLIHY-UHFFFAOYSA-N | |
| 72812 | |
| 4,7-diphenyl-1,10-phenanthroline |
| C24H16N2 | |
| MFCD00004976 | |
| bathophenanthroline, 1,10-phenanthroline, 4,7-diphenyl, bphen, 1,10-bathophenanthroline, bathophenanthrolin, bathophenanthrolin german, unii-4a2b091f0g, 4,7-diphenyl-1,10-diazaphenanthrene, gnf-pf-4554, 4,7-diphenylpyridino 3,2-h quinoline | |
| CHEBI:77995 | |
| C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5 |
Safety and Handling
RTECSNumber : SF8427000
TSCA : Yes