missing translation for 'onlineSavingsMsg'
Learn More

Bathophenanthroline, For Spectrophotometric Det. of Fe in Serum, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111013313
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013313 1 g
111013314 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013313 Supplier Merck Emd Millipore Supplier No. 118801GF

Bathophenanthroline (BPhen) is a chelating agent and is specific for ferrous ions.

Chemical Identifiers

MDL Number MFCD00004976
Synonym 4,7-Diphenyl-1,10-phenanthroline; BPhen

Specifications

Melting Point 218°C to 220°C; 218°C to 222°C
CAS 1662-01-7
Linear Formula C24H16N2
Quantity 1 g
Formula Weight 332.40
Percent Purity ≥99.0% (NT); ≥99.0%
RTECSNumber SF8427000