Learn More
Bathophenanthroline, 98+%
CAS: 1662-01-7 | C24H16N2 | 332.406 g/mol
Supplier: Thermo Scientific Chemicals A1425806
Description
Bathophenanthroline is widely used for determination of iron in serum and urine by colorimtetry. It acts as buffer layer to improve the efficiency of organic photovoltaic cells. It is very useful as a scavenger reagent, for the removal of traces of iron and copper from reagent solutions, which is used in the trace metal determination. Iron bathophenanthroline complex is involved as a redox mediating agent for imaging surface immobilized deoxyribonucleic acid (DNA) by using Scanning Electrochemical Microscopy (SECM).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBathophenanthroline is widely used for determination of iron in serum and urine by colorimtetry. It acts as buffer layer to improve the efficiency of organic photovoltaic cells. It is very useful as a scavenger reagent, for the removal of traces of iron and copper from reagent solutions, which is used in the trace metal determination. Iron bathophenanthroline complex is involved as a redox mediating agent for imaging surface immobilized deoxyribonucleic acid (DNA) by using Scanning Electrochemical Microscopy (SECM).
Solubility
Soluble in organic solvents. Slightly soluble in acidic aqueous solutions. Insoluble in neutral or alkaline solutions.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications
| Bathophenanthroline | |
| 218°C to 222°C | |
| MFCD00004976 | |
| 261048 | |
| Soluble in organic solvents. Slightly soluble in acidic aqueous solutions. Insoluble in neutral or alkaline solutions. | |
| C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5 | |
| 332.406 | |
| CHEBI:77995 | |
| ≥98% |
| 1662-01-7 | |
| C24H16N2 | |
| 5 g | |
| bathophenanthroline, 1,10-phenanthroline, 4,7-diphenyl, bphen, 1,10-bathophenanthroline, bathophenanthrolin, bathophenanthrolin german, unii-4a2b091f0g, 4,7-diphenyl-1,10-diazaphenanthrene, gnf-pf-4554, 4,7-diphenylpyridino 3,2-h quinoline | |
| DHDHJYNTEFLIHY-UHFFFAOYSA-N | |
| 4,7-diphenyl-1,10-phenanthroline | |
| 72812 | |
| 332.41 |
Chemical Identifiers
| 1662-01-7 | |
| 332.406 | |
| DHDHJYNTEFLIHY-UHFFFAOYSA-N | |
| 72812 | |
| 4,7-diphenyl-1,10-phenanthroline |
| C24H16N2 | |
| MFCD00004976 | |
| bathophenanthroline, 1,10-phenanthroline, 4,7-diphenyl, bphen, 1,10-bathophenanthroline, bathophenanthrolin, bathophenanthrolin german, unii-4a2b091f0g, 4,7-diphenyl-1,10-diazaphenanthrene, gnf-pf-4554, 4,7-diphenylpyridino 3,2-h quinoline | |
| CHEBI:77995 | |
| C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5 |
Safety and Handling
EINECSNumber : 216-767-1
RTECSNumber : SF8427000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only