missing translation for 'onlineSavingsMsg'
Learn More

Bathophenanthroline, 98+%

Catalog No. AAA1425803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1425803 1 g
AAA1425806 5 g
AAA1425814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1425803 Supplier Thermo Scientific Chemicals Supplier No. A1425803

CAS: 1662-01-7 | C24H16N2 | 332.406 g/mol

Bathophenanthroline is widely used for determination of iron in serum and urine by colorimtetry. It acts as buffer layer to improve the efficiency of organic photovoltaic cells. It is very useful as a scavenger reagent, for the removal of traces of iron and copper from reagent solutions, which is used in the trace metal determination. Iron bathophenanthroline complex is involved as a redox mediating agent for imaging surface immobilized deoxyribonucleic acid (DNA) by using Scanning Electrochemical Microscopy (SECM).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bathophenanthroline is widely used for determination of iron in serum and urine by colorimtetry. It acts as buffer layer to improve the efficiency of organic photovoltaic cells. It is very useful as a scavenger reagent, for the removal of traces of iron and copper from reagent solutions, which is used in the trace metal determination. Iron bathophenanthroline complex is involved as a redox mediating agent for imaging surface immobilized deoxyribonucleic acid (DNA) by using Scanning Electrochemical Microscopy (SECM).

Solubility
Soluble in organic solvents. Slightly soluble in acidic aqueous solutions. Insoluble in neutral or alkaline solutions.

Notes
Store in cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1662-01-7
Molecular Formula C24H16N2
Molecular Weight (g/mol) 332.406
MDL Number MFCD00004976
InChI Key DHDHJYNTEFLIHY-UHFFFAOYSA-N
Synonym bathophenanthroline, 1,10-phenanthroline, 4,7-diphenyl, bphen, 1,10-bathophenanthroline, bathophenanthrolin, bathophenanthrolin german, unii-4a2b091f0g, 4,7-diphenyl-1,10-diazaphenanthrene, gnf-pf-4554, 4,7-diphenylpyridino 3,2-h quinoline
PubChem CID 72812
ChEBI CHEBI:77995
IUPAC Name 4,7-diphenyl-1,10-phenanthroline
SMILES C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5

Specifications

Melting Point 218°C to 222°C
Quantity 1 g
Beilstein 261048
Solubility Information Soluble in organic solvents. Slightly soluble in acidic aqueous solutions. Insoluble in neutral or alkaline solutions.
Formula Weight 332.41
Percent Purity ≥98%
Chemical Name or Material Bathophenanthroline
EINECSNumber 216-767-1
RTECSNumber SF8427000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only