missing translation for 'onlineSavingsMsg'
Learn More

Bathocuproinedisulfonic acid, disodium salt hydrate, 97%

Catalog No. AC164060010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164060010 1 g Glass bottle
AC164060050 5 g Glass bottle
AC164060250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC164060010 Supplier Thermo Scientific Chemicals Supplier No. 164060010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Bathocuproinedisulfonic acid, disodium salt hydrate, 97%, CAS Number-1257642-74-2| C26H18N2Na2O6S2 xH2O | 564.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1257642-74-2
Molecular Formula C26H18N2Na2O6S2
Molecular Weight (g/mol) 564.54
MDL Number MFCD00149974
InChI Key RNGKZLRAVYPLJC-UHFFFAOYSA-L
Synonym disodium bathocuproinedisulfonate, sodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, bathocuproinedisulfonic acid sodium salt, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate disodium salt, dipotassium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, disodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate
PubChem CID 15678335
IUPAC Name disodium;2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate
SMILES [Na+].[Na+].CC1=NC2=C(C=CC3=C2N=C(C)C(=C3C2=CC=CC=C2)S([O-])(=O)=O)C(C2=CC=CC=C2)=C1S([O-])(=O)=O

Specifications

Melting Point 300.0°C
Assay Percent Range disulfonate) (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble in water
Formula Weight 564.54
Percent Purity 97%
Chemical Name or Material Bathocuproinedisulfonic acid, disodium salt hydrate
EINECSNumber 258-111-7

RUO – Research Use Only