missing translation for 'onlineSavingsMsg'
Learn More

Bathocuproine (purified by sublimation) 99.0+%, TCI America™

Supplier:  TCI America B26945G

 View more versions of this product

Catalog No. B26945G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Bathocuproine (purified by sublimation)
283°C
MFCD00004972
bathocuproine, bathocuproin, 1,10-phenanthroline, 2,9-dimethyl-4,7-diphenyl, unii-9thp2v94fx, 9thp2v94fx, 2,9-dimethyl-4,7-diphenylpyridino 3,2-h quinoline, pubchem19657, bathocuproin 1g, bidd:gt0563
CC1=CC(C2=CC=CC=C2)=C2C=CC3=C(C=C(C)N=C3C2=N1)C1=CC=CC=C1
360.46
360.46
Crystalline Powder
4733-39-5
C26H20N2
5 g
STTGYIUESPWXOW-UHFFFAOYSA-N
2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline
65149
≥99.0% (HPLC,T)

Chemical Identifiers

4733-39-5
360.46
STTGYIUESPWXOW-UHFFFAOYSA-N
65149
CC1=CC(C2=CC=CC=C2)=C2C=CC3=C(C=C(C)N=C3C2=N1)C1=CC=CC=C1
C26H20N2
MFCD00004972
bathocuproine, bathocuproin, 1,10-phenanthroline, 2,9-dimethyl-4,7-diphenyl, unii-9thp2v94fx, 9thp2v94fx, 2,9-dimethyl-4,7-diphenylpyridino 3,2-h quinoline, pubchem19657, bathocuproin 1g, bidd:gt0563
2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-1154

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions