missing translation for 'onlineSavingsMsg'
Learn More

Bathocuproine (purified by sublimation) 99.0+%, TCI America™

Catalog No. B26941G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26941G 1 g
B26945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B26941G Supplier TCI America Supplier No. B26941G

Chemical Identifiers

CAS 4733-39-5
Molecular Formula C26H20N2
Molecular Weight (g/mol) 360.46
MDL Number MFCD00004972
InChI Key STTGYIUESPWXOW-UHFFFAOYSA-N
Synonym bathocuproine, bathocuproin, 1,10-phenanthroline, 2,9-dimethyl-4,7-diphenyl, unii-9thp2v94fx, 9thp2v94fx, 2,9-dimethyl-4,7-diphenylpyridino 3,2-h quinoline, pubchem19657, bathocuproin 1g, bidd:gt0563
PubChem CID 65149
IUPAC Name 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline
SMILES CC1=CC(C2=CC=CC=C2)=C2C=CC3=C(C=C(C)N=C3C2=N1)C1=CC=CC=C1

Specifications

Melting Point 283°C
Quantity 1 g
Formula Weight 360.46
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bathocuproine (purified by sublimation)
EINECSNumber (5)-1154
TSCA Yes